Orders LinkContactUsLinkComponent
Scoops
Scoops
400 mm, 160 mm, White
Catalog # SCRW4280400
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # SCRW4280400
Supplier:  SCHROEDER WILESCO
Scoops
Scoops
400 mm, 160 mm, White
Price
9,80 €
Log in for availability
1 * 1 items
Product Details & Documents
These coloured scoops made from PP are suitable for food contact.
Specifications
  • Width:
    160 mm
  • Colour:
    White
  • Length:
    400 mm
  • Seq:
    05