Process wipes, KIMTECH PREP*
Roll, Blue, 380×340 mm, 1 roll of 500 sheets
Catalog # 115-2222
Supplier: KIMBERLY CLARK
Jump to:
Product Details & Documents
Strong, low-lint wipes for repeat use cleaning with chemicals and solvents. Perfect for printing, pre and post-paint applications, glass and appliance manufacturing, engine maintenance.
- PP, reusable
- Great for wiping oils and grease
- Acid, base, and solvent resistant
- Roll 115-2222 is compatible with dispensers 115-0045, 115-1533 and 115-2038
Colour: Blue
Specifications
- Packed:1 roll of 500 sheets
- Colour:Blue
- Description:Roll
- L×W:380×340 mm