Measuring scoops
500 ml, 350 mm
Catalog # 231-0331
Supplier: Brand
Jump to:
Product Details & Documents
HDPE, natural coloured. With flat, bevelled end.
- Good chemical resistance
- Good thermal resistance
- Robust, unbreakable construction
Specifications
- Capacity:500 ml
- Length:350 mm
- Seq:02
- Cat. No.:231-0331