Orders LinkContactUsLinkComponent
Spray bottles
Spray bottles
850 ml
Catalog # 215-0110
Supplier:   Brand
Spray bottles
Catalog # 215-0110
Supplier:  Brand
Spray bottles
Spray bottles
850 ml
Price
59,80 €
Log in for availability
1 * 5 items
Product Details & Documents
HDPE, white.
  • Good chemical resistance
  • Good temperature resistance
  • Robust, convenient, stable
By turning the nozzle, delivery can be varied from a fine spray to a solid stream.
Specifications
  • Capacity:
    850 ml
  • Seq:
    03