ContactUsLinkComponent
EK 50, SPRING CLAMP, FOR LABORATORY FLASKS UP TO DIA. 52 MM, FOR EASY AND SAFE FIXING OF FLASKS ON THE BOTTOM OF INSERT BASKETS OR U TENSIL HOLDERS WITH MESH SIZES UP TO 12,5 X 12,5 MM 1 * 1 items
Catalog # BAND055
undefined
EK 50, SPRING CLAMP, FOR LABORATORY FLASKS UP TO DIA. 52 MM, FOR EASY AND SAFE FIXING OF FLASKS ON THE BOTTOM OF INSERT BASKETS OR U TENSIL HOLDERS WITH MESH SIZES UP TO 12,5 X 12,5 MM 1 * 1 items
Catalog # BAND055
Frequently Bought Together