Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Purple
  WHEAW242417-A
Dropper tips, WHEATON®
  WHEAW242417-A
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Purple
471,00 €
1 * 1.000 items
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
 : Tips available in different colours and packaged in case of 100 or 1000.
  • Colour:
    Purple
  • Seq:
    36