Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Orange
  215-4824
Dropper tips, WHEATON®
  215-4824
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Orange
471,00 €
1 * 1.000 items
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
 : Tips available in different colours and packaged in case of 100 or 1000.
  • Colour:
    Orange
  • Seq:
    28