Dropper tips, WHEATON®
Pink
WHEAW242421-A
LDPE, coloured.
- Extended controlled dropper tips
- Dispense one drop at a time
- Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
- Colour:Pink
- Seq:43