Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Yellow
  215-4819
Dropper tips, WHEATON®
  215-4819
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Yellow
360,00 €
1 * 1.000 items
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
  • Colour:
    Yellow
  • Seq:
    18