Dropper tips, WHEATON®
Orange
215-4833
LDPE, coloured.
- Extended controlled dropper tips
- Dispense one drop at a time
- Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
- Colour:Orange
- Seq:67