Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Grey
Catalog # 215-4780
Supplier:   DWK Life Sciences
Dropper tips, WHEATON®
Catalog # 215-4780
Supplier:  DWK Life Sciences
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Grey
Price
41,40 €
Log in for availability
1 * 100 items
Product Details & Documents
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
Specifications
  • Colour:
    Grey
  • Seq:
    11