Dropper tips, WHEATON®
Green
215-4807
LDPE, coloured.
- Extended controlled dropper tips
- Dispense one drop at a time
- Average drop size is 44 ±6 μl
: Tips available in different colours and packaged in case of 100 or 1000.
- Colour:Green
- Seq:70
LDPE, coloured.
: Tips available in different colours and packaged in case of 100 or 1000.