Dropper tip caps, WHEATON®
20-410, Orange
Catalog # 215-4022
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifications
- Thread:20-410
- Seq:39
- Cap colour:Orange
- Cat. No.:215-4022