Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
1000 ml, 400 mm
Catalog # 231-0071
Supplier:   KARTELL
Measuring scoops
Catalog # 231-0071
Supplier:  KARTELL
Measuring scoops
Measuring scoops
1000 ml, 400 mm
Price
179,00 €
Log in for availability
1 * 6 items
Product Details & Documents
PP, white. With flat, bevelled end and reinforced handle.
  • Excellent chemical resistance, unbreakable
  • Heat resistant up to 130 °C, autoclavable
  • The capacity of the scoop is stamped into the handle for easy reading
Specifications
  • Capacity:
    1000 ml
  • Length:
    400 mm
  • Seq:
    14