Orders LinkContactUsLinkComponent
Separating funnels
Separating funnels
100 ml, 14/23 NS
Catalog # QVFLALG0050702
Supplier:   QVF LABORTECHNIK
Separating funnels
Catalog # QVFLALG0050702
Supplier:  QVF LABORTECHNIK
Separating funnels
100 ml, 14/23 NS
Price
125,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Made of borosilicate glass.
  • Stopcock with glass key
  • Without graduations
  • Excellent chemical and temperature resistance
The separating funnels are used for discontinuous shaking out of solvents.
Certifications: Complies to DIN 12 451.
Specifications
  • Socket size:
    14/23 NS
  • Capacity:
    100 ml
  • Seq:
    02