Orders LinkContactUsLinkComponent
Mop systems, Edgeless®
Standard edgeless mop head, non sterile, -, -, -
  115-0174
 :   CONTEC
Mop systems, Edgeless®
  115-0174
 :  CONTEC
Mop systems, Edgeless®
Mop systems, Edgeless®
Standard edgeless mop head, non sterile, -, -, -
62,00 €
1 * 1 items
Designed for wet mopping in controlled environments and cleanrooms for effective cleaning and retrieval of excess fluid. Ideal for use in sterile environments. Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption.
  • Mop heads are free from any cellulose contaminants
  • Stainless bucket, downpress wringer and insert maximise wringing pressure and effectiveness
  • All system components and buckets are compatible with gamma, ETO, and autoclave sterilisation
Mop heads are laundered and packaged in a ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
 : All mop system components must be ordered separately. Plastic bucket systems include cart, buckets, wringer, and casters. Stainless steel bucket systems include trolley, buckets, wringer, and casters.
  • Capacity:
    -
  • Description:
    Standard edgeless mop head, non sterile
  • Dimensions:
    -
  • Length:
    -
  • Seq:
    02