Orders LinkContactUsLinkComponent
Sample Bags without Closure
Sample Bags without Closure
1100×700 mm, 0,05 mm
Catalog # SCAC2047
Sample Bags without Closure
Catalog # SCAC2047
Sample Bags without Closure
Sample Bags without Closure
1100×700 mm, 0,05 mm
Price
Inquire For Price
Log in for availability
1 * 250 items
Product Details & Documents
Bags made from PE transparent, for the storage or transport of samples.
Available in several sizes.
Caution: Supplied without closure ties.
Specifications
  • L×W:
    1100×700 mm
  • Seq:
    19
  • Thickness:
    0,05 mm