Orders LinkContactUsLinkComponent
Assortment Boxes, PS
Assortment box, with 12 compartments, Transparent, 210×120×36 mm, PS
Catalog # HUEN616300
Supplier:   hünersdorff GmbH
Assortment Boxes, PS
Catalog # HUEN616300
Supplier:  hünersdorff GmbH
Assortment Boxes, PS
Assortment Boxes, PS
Assortment box, with 12 compartments, Transparent, 210×120×36 mm, PS
Price
10,70 €
Log in for availability
1 * 1 items
Product Details & Documents
These assortment boxes made of high impact strength polystyrene (PS) are suitable for packaging purposes and create order in workshops and in household.
  • Available either with versatile compartment subdivisions and versions or without compartments
  • Transparent lid and bottom for optimum display of the contents or in colours suitable for foods
  • Printing possible on request
Specifications
  • Colour:
    Transparent
  • Description:
    Assortment box, with 12 compartments
  • Material:
    PS
  • Seq:
    04
  • W×D×H:
    210×120×36 mm