Orders LinkContactUsLinkComponent
Assortment Boxes, PS
Assortment box, with 6 compartments, Transparent, 335×225×55 mm, PS
  HUEN612200
Assortment Boxes, PS
  HUEN612200
Assortment Boxes, PS
Assortment Boxes, PS
Assortment box, with 6 compartments, Transparent, 335×225×55 mm, PS
18,60 €
1 * 1 items
These assortment boxes made of high impact strength polystyrene (PS) are suitable for packaging purposes and create order in workshops and in household.
  • Available either with versatile compartment subdivisions and versions or without compartments
  • Transparent lid and bottom for optimum display of the contents or in colours suitable for foods
  • Printing possible on request
  • Colour:
    Transparent
  • Description:
    Assortment box, with 6 compartments
  • Material:
    PS
  • Seq:
    12
  • W×D×H:
    335×225×55 mm