Orders LinkContactUsLinkComponent
Gouges, Aesculap®
Gouges, Aesculap®
Gouge, inclined forward, Wagner, 9 mm, 340 mm
Catalog # AESCND120R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCND120R
Supplier:  Aesculap
Gouges, Aesculap®
Gouge, inclined forward, Wagner, 9 mm, 340 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Type:
    Wagner
  • Width:
    9 mm
  • Description:
    Gouge, inclined forward
  • Length:
    340 mm
  • Seq:
    45