Gouges, Aesculap®
Lamina gouge, cranked, Capener, 6 mm, 235 mm
Catalog # AESCFL437R
Supplier: Aesculap
Jump to:
Product Details & Documents
Specifications
- Type:Capener
- Width:6 mm
- Description:Lamina gouge, cranked
- Length:235 mm
- Seq:50