Gouges, Aesculap®
Gouge, Lexer, 25 mm, 220 mm
Catalog # AESCFL417R
Supplier: Aesculap
Jump to:
Product Details & Documents
Specifications
- Type:Lexer
- Width:25 mm
- Description:Gouge
- Length:220 mm
- Seq:14