Orders LinkContactUsLinkComponent
Gouges, Aesculap®
Gouges, Aesculap®
Gouge, Partsch, 5 mm, 170 mm
Catalog # AESCFL225R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL225R
Supplier:  Aesculap
Gouges, Aesculap®
Gouge, Partsch, 5 mm, 170 mm
Price
65,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Type:
    Partsch
  • Width:
    5 mm
  • Description:
    Gouge
  • Length:
    170 mm
  • Seq:
    29