Orders LinkContactUsLinkComponent
Gouges, Aesculap®
Gouges, Aesculap®
Gouge, -, 6 mm, 140 mm
Catalog # AESCFL132R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL132R
Supplier:  Aesculap
Gouges, Aesculap®
Gouge, -, 6 mm, 140 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Type:
    -
  • Width:
    6 mm
  • Description:
    Gouge
  • Length:
    140 mm
  • Seq:
    02