Orders LinkContactUsLinkComponent
Dissecting needles
Dissecting needles
Bent, 140 mm
Catalog # 233-0105
Supplier:   BOCHEM
Dissecting needles
Catalog # 233-0105
Supplier:  BOCHEM
Dissecting needles
Dissecting needles
Bent, 140 mm
Price
7,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Dissecting needles completely made of ¹⁸/₁₀ stainless steel.
Specifications
  • Form:
    Bent
  • Length:
    140 mm