Orders LinkContactUsLinkComponent
Double spatulas, flexible
Double spatulas, flexible
210 mm, 48 mm, 10 mm
Catalog # BOCH3623
Supplier:   BOCHEM
Double spatulas, flexible
Catalog # BOCH3623
Supplier:  BOCHEM
Double spatulas, flexible
Double spatulas, flexible
210 mm, 48 mm, 10 mm
Price
4,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 18/10.
Specifications
  • Length:
    210 mm
  • Blade width:
    10 mm
  • Blade length:
    48 mm