Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides with epoxy coating, Black, 10, 6 mm
Catalog # 631-0457
Supplier:   Epredia
Cavity slides
Catalog # 631-0457
Supplier:  Epredia
Cavity slides
Cavity slides
Cavity slides with epoxy coating, Black, 10, 6 mm
Price
129,00 €
Log in for availability
1 * 100 items
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
  • Adhesive surface and hydrophobic mask
  • Hydrophobic epoxy- or PTFE-coating
  • (*) with rings or wells
Developped for clinical diagnosis applications.
Ordering information: Available with rings or wells (*).
Specifications
  • No. of wells:
    10
  • Colour:
    Black
  • Description:
    Cavity slides with epoxy coating
  • Seq:
    010
  • Well Ø:
    6 mm