Cover glasses, #1
22×32 mm, #1, Borosilicate glass
Supplier: MARIENFELD
Jump to:
Product Details & Documents
These cover glasses are suitable for fluorescence microscopy and in vitro diagnostic (IVD) applications.
- Long storage, non corroding
- Made of clear borosilicate D 263™ M glass, Hydrolytic Class I
- Do not stick together
- Excellent flat, smooth surfaces
Certifications: IVD 98/79/EC.
Packaging: In plastic boxes with hinged lid – 1000 pieces per box.
Documents
Certificates
Search by Lot NumberSpecifications
- No.:#1
- L×W:22×32 mm
- Material:Borosilicate glass