Orders LinkContactUsLinkComponent
Dropper tips, WHEATON®
Blue
  215-4797
Dropper tips, WHEATON®
  215-4797
Dropper tips, WHEATON®
Dropper tips, WHEATON®
Blue
54,20 €
1 * 100 items
LDPE, coloured.
  • Extended controlled dropper tips
  • Dispense one drop at a time
  • Average drop size is 44 ±6 μl
Ordering information: Tips available in different colours and packaged in case of 100 or 1000.
  • Colour:
    Blue
  • Seq:
    49