Polyethylene stoppers
White, PE, 29/32
AZLOTRF597
These PE stoppers are ideally suited for use with glass or plastic volumetric flasks and measuring cylinders etc. which have longer 'B' length sockets.
Large head provides a good grip and gives protection if glassware is accidently knocked over.
: 10/package
- NS:29/32
- Colour:White
- Material:PE