Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
100 ml, 200 mm
Catalog # 231-0337
Supplier:   Brand
Measuring scoops
Catalog # 231-0337
Supplier:  Brand
Measuring scoops
Measuring scoops
100 ml, 200 mm
Price
24,60 €
Log in for availability
1 * 12 items
Product Details & Documents
PP, white. With flat, bevelled end and reinforced handle.
  • Excellent chemical resistance, unbreakable
  • The capacity of the scoop is stamped into the handle for easy reading
Specifications
  • Capacity:
    100 ml
  • Length:
    200 mm