Wire triangles
With smooth earthenware pipes, tube length 80 mm
Catalog # HECH41065008
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
These wire triangles with plain clay tubes are made of galvanised steel wire.
Specifications
- Description:With smooth earthenware pipes, tube length 80 mm