Embedding cassettes, M480
Yellow
Catalog # 720-0835
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures.
- Resistant to all commonly used histological solvents
- Designed to accept standard metal lids (M481)
- Slanted writing surface (45° angle) accepts markings easily
Different colours available.
Specifications
- Colour:Yellow
- Seq:08
Accessories
Product Information
- Catalog #ColourAvailabilityPrice