Orders LinkContactUsLinkComponent
Embedding cassettes, M480
Pink
  720-0833
Embedding cassettes, M480
  720-0833
Embedding cassettes, M480
Embedding cassettes, M480
Pink
213,00 €
1 * 1.500 items
Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures.
  • Resistant to all commonly used histological solvents
  • Designed to accept standard metal lids (M481)
  • Slanted writing surface (45° angle) accepts markings easily
Different colours available.
  • Colour:
    Pink
  • Seq:
    06
Accessories
  •  
    Colour
  • 720-0403
    Silver
    1 * 25 items - 110,00 €