TIPS IRRADIATED IN BOX FOR BIO DILUTOR 1 * 60 items
Catalog # IULA1471
Supplier: IUL Instruments SA
Frequently Bought Together
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
VWR®, Cell Culture Insert Plates, PET
Supplier: VWR Collection
Freezer boxes (Cryoboxes), True North®
Supplier: HEATHROW
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Protein coated cell culture plates, CELLCOAT®, collagen type I
Supplier: Greiner Bio-One
Culture tubes, sterile, Cultubes™
Supplier: Simport Scientific
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific
Glass bottom microplates, SensoPlate™
Supplier: Greiner Bio-One
Measuring scoops
Supplier: VITLAB
Amine and carboxyl microplates, Corning® PureCoat™
Supplier: Corning
Micro Test Tubes, Graduated
Supplier: KARTELL
Screw Cap Tubes
Supplier: MEGRO