Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5ml graduated Pasteur pipette, 1.0ml div, 217mm, pk250 1 * 250 items
Catalog # 612-0012
Supplier:
Alpha Laboratories
Expand image
5ml graduated Pasteur pipette, 1.0ml div, 217mm, pk250 1 * 250 items
Catalog # 612-0012
Supplier:
Alpha Laboratories
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection