Orders LinkContactUsLinkComponent
Storage Boxes
Blue, 595×395×165 mm
Catalog # 216-2873
Supplier:   Gilac PROFESSIONNEL
Storage Boxes
Catalog # 216-2873
Storage Boxes
Storage Boxes
Blue, 595×395×165 mm
Price
109,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.
  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
Specifications
  • Colour:
    Blue
  • Seq:
    08
  • W×D×H:
    595×395×165 mm