ContactUsLinkComponent
Dms/n,universal flow-thru vessel for multiparameter measurements with up to four sensors, e.g. for use in the discharge of a waste w ater treatment plantplease order suitable sensor flange separately 1 * 1 items
  WTWO203749
Dms/n,universal flow-thru vessel for multiparameter measurements with up to four sensors, e.g. for use in the discharge of a waste w ater treatment plantplease order suitable sensor flange separately 1 * 1 items
  WTWO203749
Frequently Bought Together