Filter sets
Filter set 44, excitation BP 475/40, beam splitter FT 500, emission BP 530/50
Catalog # ZEIS0000001114459
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Description:Filter set 44, excitation BP 475/40, beam splitter FT 500, emission BP 530/50
- Seq:11