Filter sets
Filter set 31, excitation BP 565/30, beam splitter FT 585, emission BP 620/60
Catalog # ZEIS0000001031350
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Description:Filter set 31, excitation BP 565/30, beam splitter FT 585, emission BP 620/60
- Seq:07