Orders LinkContactUsLinkComponent
Test strips
Test strips
Chloride, 30 - 600  mg/L, TS, 40
Catalog # 705-0186
Supplier:   Hach
Test strips
Catalog # 705-0186
Supplier:  Hach
CAS Number:  
Test strips
Test strips
Chloride, 30 - 600  mg/L, TS, 40
CAS Number:  
Price
45,40 €
Log in for availability
1 * 40 items
Product Details & Documents
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
Documents
Specifications
Specifications
  • Pk:
    40
  • Description:
    Chloride
  • Type:
    TS
  • Range:
    30 - 600  mg/L
Specifications