Orders LinkContactUsLinkComponent
Test strips
Test strips
Copper test strips, 0 - 3 mg/L, TS, 25
Catalog # HACH2745125
Test strips
Catalog # HACH2745125
Supplier:  
CAS Number:  
Test strips
Test strips
Copper test strips, 0 - 3 mg/L, TS, 25
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Test Strips, Copper, 0 - 3 mg/l, 25 tests, Steps: 0, 0.2, 0.5, 1, 3 mg/l, 1 * 25 items Catalog #HACH2745125-EU
Product HACH2745125-EU is a replacement for HACH2745125 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
  • Portable - ideal for field testing of surface water
  • Easy to use
Documents
Specifications
Specifications
  • Pk:
    25
  • Description:
    Copper test strips
  • Type:
    TS
  • Range:
    0 - 3 mg/L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.