Test strips
Copper test strips, 0 - 3 mg/L, TS, 25
Catalog # HACH2745125
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Test Strips, Copper, 0 - 3 mg/l, 25 tests, Steps: 0, 0.2, 0.5, 1, 3 mg/l, 1 * 25 items Catalog #HACH2745125-EU
Product HACH2745125-EU is a replacement for HACH2745125 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.
- Portable - ideal for field testing of surface water
- Easy to use
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:25
- Description:Copper test strips
- Type:TS
- Range:0 - 3 mg/L