ContactUsLinkComponent
Test Strips, Nitrate/Nitrite, 0 - 50 mg/l NO3 / 0 - 3 mg/l NO2, 25 tests, Steps: 0, 1, 2, 5, 10, 20, 50 mg/l NO3, Steps: 0, 0.15, 0. 3, 1, 1.5, 3 NO2 1 * 25 items
Catalog # 705-0188
Supplier:  Hach
undefined
Test Strips, Nitrate/Nitrite, 0 - 50 mg/l NO3 / 0 - 3 mg/l NO2, 25 tests, Steps: 0, 1, 2, 5, 10, 20, 50 mg/l NO3, Steps: 0, 0.15, 0. 3, 1, 1.5, 3 NO2 1 * 25 items
Catalog # 705-0188
Supplier:  Hach
Frequently Bought Together