Orders LinkContactUsLinkComponent
Microtubes, with Thread
2,0 ml, 1000/case
Catalog # SARS72.693.005
Supplier:   SARSTEDT
Microtubes, with Thread
Catalog # SARS72.693.005
Supplier:  SARSTEDT
Microtubes, with Thread
2,0 ml, 1000/case
Price
178,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Specifications
  • Packed:
    1000/case
  • Capacity:
    2,0 ml
  • Seq:
    19