Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

pH test kits, model 17

pH, mid range, model 17H, pH 6,6 - 8,4, Phenol red, 1 KIT
  HACH147008
 :   Hach
pH test kits, model 17
  HACH147008
 :  Hach
 :  
pH test kits, model 17
pH test kits, model 17

pH, mid range, model 17H, pH 6,6 - 8,4, Phenol red, 1 KIT

 :  
 
187,00 €
   
1 * 1 KIT
 

It is a simple, low cost colour disc method which is one of the most accurate visual test kit methods for pH.

 : Kit includes indicator reagent, colour disc, comparator box, two viewing tubes, instructions and carrying case.

 

 

  • Chemical method:
    Phenol red
  • Pk:
    1 KIT
  • Description:
    pH, mid range, model 17H
  • Range:
    pH 6,6 - 8,4
  • Cat. No.:
    HACH147008

 

Accessories

 

  •  
    Description
    Pk
     
     
  • HACH173006
    Colour viewing tube, glass
    6
       
    1 * 6 items - 48,80 €

 

  •  
    Description
    Pk
     
     
  • HACH9261100
    Color disc pH, phenol red, 6,6 - 8,4
    1
       
    1 * 1 items - 112,00 €

 

  •  
    Description
    Pk
     
     
  • HACH173106
    Stoppers for glass viewing tube
    6
       
    1 * 6 items - 19,90 €

 

  •  
    Description
    Pk
     
     
  • HACH173200
    Colour comparator box
    1
       
    1 * 1 items - 23,70 €

 

  •  
    Description
    Pk
     
     
  • HACH4660004
    Colour viewing tube, plastic with cap
    4
       
    1 * 4 items - 33,10 €