Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

pH test kits, model 17

pH, wide range, model 17N, pH 4,0 - 10,0, 1 KIT
  HACH147011
 :   Hach
pH test kits, model 17
  HACH147011
 :  Hach
 :  
pH test kits, model 17
pH test kits, model 17

pH, wide range, model 17N, pH 4,0 - 10,0, 1 KIT

 :  
 
206,00 €
   
1 * 1 KIT
 

It is a simple, low cost colour disc method which is one of the most accurate visual test kit methods for pH.

 : Kit includes indicator reagent, colour disc, comparator box, two viewing tubes, instructions and carrying case.

 

 

  • Pk:
    1 KIT
  • Description:
    pH, wide range, model 17N
  • Range:
    pH 4,0 - 10,0

 

Accessories

 

  •  
    Description
    Pk
     
     
  • HACH173006
    Colour viewing tube, glass
    6
       
    1 * 6 items - 48,80 €

 

  •  
    Description
    Pk
     
     
  • HACH9261100
    Color disc pH, phenol red, 6,6 - 8,4
    1
       
    1 * 1 items - 112,00 €

 

  •  
    Description
    Pk
     
     
  • HACH173106
    Stoppers for glass viewing tube
    6
       
    1 * 6 items - 19,90 €

 

  •  
    Description
    Pk
     
     
  • HACH173200
    Colour comparator box
    1
       
    1 * 1 items - 23,70 €

 

  •  
    Description
    Pk
     
     
  • HACH4660004
    Colour viewing tube, plastic with cap
    4
       
    1 * 4 items - 33,10 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.