Emax microplate reader (90V - 240V) with current version of SoftMax Pro. Includes: Precision endpoint reading Wavelength range 400 nm - 750 nm (405, 450, 490, 650 nm*), User manual, Dust cover 10 blanking templates, 1 year MDCare PhD Technical Suppor 1 * 1 items
Catalog # MLDVEMAX
Supplier: Molecular Devices
Frequently Bought Together