Objectives
Fluar-Planachromatic objectives, N.A. of 0,25, WD of 12,5 mm
Catalog # ZEIS4401250000000
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Magnification:5×
- Description:Fluar-Planachromatic objectives, N.A. of 0,25, WD of 12,5 mm
- Seq:30