Orders LinkContactUsLinkComponent
Colour coders for cryovials, CryoColorCode, Nunc™
Red
  479-6000
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
  479-6000
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
Colour coders for cryovials, CryoColorCode, Nunc™
Red
27,60 €
1 * 500 items
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.
  • Colour:
    Red
  • Seq:
    03
  • Cat. No.:
    479-6000
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact