Orders LinkContactUsLinkComponent
Colour coders for cryovials, CryoColorCode, Nunc™
Colour coders for cryovials, CryoColorCode, Nunc™
White
Catalog # 479-2481
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
Catalog # 479-2481
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
Colour coders for cryovials, CryoColorCode, Nunc™
White
Price
27,60 €
Log in for availability
1 * 500 items
Product Details & Documents
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.
Specifications
  • Colour:
    White
  • Seq:
    01
  • Cat. No.:
    479-2481
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact